missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, ammonium salt, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
Specifications
Specifications
Melting Point | 114.0°C |
Linear Formula | CH3CO2NH4 |
Solubility Information | Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, in methanol: 7.89 g/100mL (15°C) |
Formula Weight | 77.08 |
Percent Purity | 98+% |
Grade | HPLC |
Quantity | 50 g |
Flash Point | 136°C |
Packaging | Plastic bottle |
Chemical Name or Material | Acetic acid, ammonium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'