missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid concentrate, For 1L standard solution, 1.0 M CH3COOH (1N), Honeywell Fluka™
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Linear Formula | CH3CO2H |
UN Number | UN2789 |
Beilstein | 506007 |
Formula Weight | 60.05g/mol |
Quantity | 1 Pc. |
Flash Point | 40°C (104°F) |
Packaging | Ampoule |
Chemical Name or Material | Acetic acid concentrate |
Safety and Handling
Product Identifier
- Acetic acid concentrate
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Acetic acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction