missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid concentrate, For 1L standard solution, 1.0 M CH3COOH (1N), Honeywell Fluka™

Product Code. 15615200
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

for 1L standard solution, 1.0 M CH3COOH (1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7, 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Boiling Point 117°C to 118°C
CAS Min % ≥15.0000%, ≥80.0000%
CAS Max % ≤20.0000%, ≤85.0000%
Linear Formula CH3CO2H
UN Number UN2789
Beilstein 506007
Formula Weight 60.05
Quantity 6 x 1 Ea.
Flash Point 40°C
Packaging Ampoule
Physical Form Concentrate
Chemical Name or Material Acetic acid concentrate, For 1L standard solution, 1.0 M CH3COOH (1N)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.