missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid-d, for NMR, 98% atom D, Thermo Scientific Chemicals

Product Code. 10583911
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 758-12-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 61.058
MDL Number MFCD00051050
InChI Key QTBSBXVTEAMEQO-DYCDLGHISA-N
Synonym acetic acid-d, acetic acid-d1, monodeuteroacetic acid, acetic 2h acid, acetic acid-od, o-2h acetic acid, mono-deuteroacetic acid, ch3cood
PubChem CID 2723902
IUPAC Name deuterio acetate
SMILES CC(=O)O

Specifications

Physical Form Liquid
Color Colorless
Density 1.0500g/mL
Deuterium (D) 97.5% min.
Flash Point 40°C
Packaging Glass Bottle
Refractive Index 1.3715
Specific Gravity 1.05
Water 0.4% max. (Coulometric)
Quantity 50 g
Melting Point 15.0°C to 16.0°C
Boiling Point 116.0°C to 117.0°C
Linear Formula CH3CO2D
Formula Weight 61.06
Grade NMR
Chemical Name or Material Acetic acid-d, 98 atom % D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic acid-d
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'