missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, glacial, 99.9985% (metals basis), Thermo Scientific Chemicals

Product Code. 11311598
Click to view available options
Quantity:
25 mL
100 mL
500 mL

Acetic acid is used as a solvent for purification and production of many chemical compounds eg., vinyl acetate monomer. It is a significant chemical reagent and is mainly used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as a descaling agent in households. As a food additive, it finds application as a condiment, in the pickling of vegetables and other foods and is also an acidity regulator. In analytical chemistry, for the estimation of weakly alkaline substances such as organic amides, glacial acetic acid is used. Acetic acid is used as antimicrobial preservative or acidulates in a variety of food products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16.6°C
Boiling Point 118.1°C
UN Number UN2789
Merck Index 14,55
Solubility Information Miscible with water,ethanol,acetone,toluene and immiscible with carbon disulphide.
Formula Weight 60.05
Percent Purity 99.9985%
Density 1.049 g/mL
Quantity 25 mL
Flash Point 40°C (104°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.3721
Assay (metals basis)
Chemical Name or Material Acetic acid, Glacial
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic acid, glacial
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Acetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.