missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial (Optima™), Fisher Chemical™

Product Code. 11904901 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
FEP Bottle
Unit Size:
1 L
250mL
500mL
This item is not returnable. View return policy

Product Code. 11904901

Brand: Fisher Chemical A465500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
  • Certificate of Analysis included with each bottle.
  • Ideal for analysis by ICP-MS and high-resolution ICP-MS
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Linear Formula CH3COOH
UN Number 2789
Merck Index 15, 54
Formula Weight 60.05
Grade OPTIMA
Density 1.048g/cm³
Quantity 500 mL
Packaging FEP Bottle
Physical Form Liquid
Vapor Density 2.1
Vapor Pressure 1.52kPa at 20°C
Chemical Name or Material Acetic Acid
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Flammable liquid Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.