missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial (Optima™), Fisher Chemical™

Product Code. 11984891
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
FEP Bottle
  • Certificate of Analysis included with each bottle.
  • Ideal for analysis by ICP-MS and high-resolution ICP-MS
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Formula Weight 60.05g/mol
Quantity 1 L
Physical Form Liquid
Vapor Density 2.1
Vapor Pressure 1.52kPa at 20°C
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Flammable liquid Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'