missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid Glacial, for Trace Metal Analysis, Primar Plus™, Fisher Chemical™

£109.00 - £339.00

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10120273
View Documents
Fisher Chemical
A/0411/PB08
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10334193
View Documents
Fisher Chemical
A/0411/PB15
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10232953
View Documents
Fisher Chemical
A/0411/PB17
2.5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
CC(O)=O
C2H4O2
MFCD00036152
acetic acid

Specifications

2.5
16°C
Colorless
MFCD00036152
CC(O)=O
60.05
HDPE plastic bottle
2.1
64-19-7
117°C
C2H4O2
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
60.05g/mol
Liquid
1.52kPa at 20°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Acetic Acid

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.