missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, potassium salt, 97%, pure, anhydrous, Thermo Scientific Chemicals

£24.82 - £65.93

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.14
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10570594
View Documents
Thermo Scientific Acros
149605000
500 g Plastic bottle
£24.82
500g
Please to purchase this item. Need a web account? Register with us today!
10091480
View Documents
Thermo Scientific Acros
149600010
1 kg Plastic bottle
£38.69
1kg
Please to purchase this item. Need a web account? Register with us today!
10388653
View Documents
Thermo Scientific Acros
149600025
2.5 kg Plastic bottle
£65.93
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

127-08-2
98.14
SCVFZCLFOSHCOH-UHFFFAOYSA-M
517044
potassium;acetate
C2H3KO2
MFCD00012458
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CHEBI:32029
CC(=O)[O-].[K+]

Specifications

292.0°C
White
100.0
97%
CH3CO2K
02, 96
>250°C
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium;acetate
517044
98.14
Pure
Acetic acid, potassium salt
127-08-2
96.0
Granular Crystalline Powder
C2H3KO2
MFCD00012458
01,906; 06,475
15, 7726
Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether
CC(=O)[O-].[K+]
98.14
CHEBI:32029
97%
Authentic
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 204-822-2

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only