missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, potassium salt, 99+%, for molecular biology, Thermo Scientific Chemicals

£30.05 - £122.61

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10163114
View Documents
Thermo Scientific Acros
397121000
100 g Glass bottle
£30.05
100g
Please to purchase this item. Need a web account? Register with us today!
10254644
View Documents
Thermo Scientific Acros
397122500
250 g Glass bottle
£41.61
250g
Please to purchase this item. Need a web account? Register with us today!
10103074
View Documents
Thermo Scientific Acros
397120010
1 kg Glass bottle
£122.61
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for molecular biology
Specifications

Chemical Identifiers

127-08-2
98.142
SCVFZCLFOSHCOH-UHFFFAOYSA-M
517044
potassium;acetate
C2H3KO2
MFCD00012458
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CHEBI:32029
CC(=O)[O-].[K+]

Specifications

127-08-2
99.0
White to Brown
C2H3KO2
MFCD00012458
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium;acetate
517044
98.14
Molecular Biology
Low Melting Solid
292.0°C
100.0
99+%
CH3CO2K
15, 7726
Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether
CC(=O)[O-].[K+]
98.142
CHEBI:32029
≥99%
>250°C
Acetic acid, potassium salt
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Acetic acid, potassium salt

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only