missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, potassium salt, 99+%, pure, anhydrous, Thermo Scientific Chemicals

Product Code. 10474985
Click to view available options
Quantity:
100 g
1 kg
5 kg
25 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.14
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Melting Point 292.0°C
Color White
CAS Min % 99.0
CAS Max % 100.0
Physical Form Granular Crystalline Powder
Quantity 25 kg
Assay Percent Range 99+%
Linear Formula CH3CO2K
Beilstein 02,96
Fieser 01,906; 06,475
Flash Point >250°C
Merck Index 15,7726
Solubility Information Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether
Formula Weight 98.14
Percent Purity ≥99%
Grade Pure
Infrared Spectrum Authentic
Chemical Name or Material Acetic acid, potassium salt, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'