missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, Reagent Grade, ≥99.0%, Honeywell™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
5 L
Packaging:
Poly Bottle
Beschreibung
- ≥95% for general laboratory use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spezifikation
Melting Point | 16.2°C (literature) |
Boiling Point | 118°C |
CAS Max % | 100.0000% |
Assay Percent Range | 99.0 to 100.5% |
Linear Formula | CH3CO2H |
UN Number | UN2789 |
Beilstein | 506007 |
Formula Weight | 60.05 |
Grade | Reagent |
Density | 1.049 g/mL at 25 °C(lit.) |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'