missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, ReagentPlus™, Honeywell
Reagent Grade,≥99%
£10.87 - £56.63
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15667660
|
Honeywell Research Chemicals
A6283-100ML |
100 mL | N/A | ||||||
15697660
|
Honeywell Research Chemicals
A6283-500ML |
500 mL | N/A | ||||||
15677660
|
Honeywell Research Chemicals
A6283-1L |
1 L | N/A | ||||||
15687660
|
Honeywell Research Chemicals
A6283-2.5L |
2.5 L | N/A | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Specifications
64-19-7 | |
CH3CO2H | |
UN2789 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
≥99% | |
1.049g/mL at 25°C | |
40°C (104°F) | |
2.07 (vs air) |
C2H4O2 | |
MFCD00036152 | |
506007 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05g/mol | |
ReagentPlus™ | |
16% at 92°F, 4% at 59°F | |
800°F | |
Acetic acid |
Safety and Handling
P280-P305 + P351 + P338-P310
missing translation for 'rtecsNumber' : AF1225000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title