missing translation for 'onlineSavingsMsg'
Learn More
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals Product Code.: 13478608

Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals

Product Code. 13478608
250 g, Plastic bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 13478608

Brand: Thermo Scientific Acros 446882500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifications

Melting Point 324.0°C
Flash Point >250°C
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH3CO2Na
Quantity 250 g
Merck Index 15,8709
Solubility Information Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name sodium acetate
Formula Weight 82.03
Percent Purity 99%
Grade HPLC
Insoluble Matter 0.005% max
Chemical Name or Material Acetic acid, sodium salt
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468757 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number446880500,446882500
Lot NumberA0457635 Certificate TypeCertificate of Analysis Date08/11/2023 Catalog Number446880500,446882500
Lot NumberA0440101 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number446880500,446882500
Lot NumberA0453232 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number446880500,446882500
Lot NumberA0400415 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number446880500,446882500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.