missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, sodium salt, 99+%, for mol biology, anhydrous, DNAse, RNAse and P, Thermo Scientific Chemicals

Product Code. 10546941
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifications

Absorbance (0.1 M in water), (0.1 M in water) at 0nm, 0.01 max. at 260nm, 0.01 max. at 280nm
pH 7 to 9.2 (5% soln. at 25°C)
CAS Min % 99.0
CAS Max % 100.0
Color White
Assay Percent Range 99+%
Linear Formula CH3CO2Na
IUPAC Name sodium acetate
Formula Weight 82.03
Percent Purity ≥99.0%
Grade Molecular Biology
Quantity 1 kg
Infrared Spectrum Authentic
Loss on Drying 1% max. (120°C)
Packaging Plastic bottle
Physical Form Crystalline Solid
Chemical Name or Material Acetic acid, sodium salt, Anhydrous, DNAse, RNAse and Protease Free
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.