missing translation for 'onlineSavingsMsg'
Learn More
Acetic acid, sodium salt trihydrate, 99+%, extra pure, Thermo Scientific Chemicals Product Code.: 10687963

Acetic acid, sodium salt trihydrate, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10687963
5 kg, Plastic drum
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10687963

Brand: Thermo Scientific Acros 123240050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3, 6131-90-4
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifications

Melting Point 58.0°C
CAS Min % 99.0
CAS Max % 100.0
Color White
Assay Percent Range 99+%
Linear Formula CH3COONa·3H2O
Beilstein 02, 96
Fieser 01,1024; 05,591
Merck Index 15, 8709
Solubility Information Solubility in water: 762g/L (20 °C). Other solubilities: 1g/19mL alcohol
IUPAC Name sodium acetate
Formula Weight 136.08
Percent Purity ≥99%
Grade Extra Pure
Quantity 5 kg
Infrared Spectrum Authentic
Packaging Plastic drum
Physical Form Adhering Crystals
Chemical Name or Material Acetic acid, sodium salt trihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467552 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number123240010,123240050
Lot NumberA0457852 Certificate TypeCertificate of Analysis Date29/11/2023 Catalog Number123240010,123240050
Lot NumberA0449522 Certificate TypeCertificate of Analysis Date26/01/2023 Catalog Number123240010,123240050
Lot NumberA0411153 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number123240010,123240050
Lot NumberA0441566 Certificate TypeCertificate of Analysis Date28/04/2022 Catalog Number123240010,123240050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetic acid, sodium salt trihydrate, 99+%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.