missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid solution EU Pharmacopoeia, Fisher Chemical™

Product Code. 12990834
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Concentration 0.1M
CAS 64-19-7
Packaging Bottle
Quantity 1000 mL
Molecular Formula C2H4O2
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(O)=O
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.05
PubChem CID 176
ChEBI CHEBI:15366
Grade Eu. Ph. Reference
Chemical Name or Material Acetic Acid Solution
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'