missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Anhydride 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
Specifications
Specifications
Color | Colorless |
Density | 1.087g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 99+% |
Packaging | Glass bottle |
Linear Formula | (CH3CO)2O |
Refractive Index | 1.3890 to 1.3910 (20°C, 589nm) |
Quantity | 1 L |
Beilstein | 02, 166 |
Fieser | 01,3; 02,7; 05,3; 06,1; 07,1; 08,1; 09,1; 11,1; 12,1; 13,93; 14,150; 15,1 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467302 | Certificate of Analysis | 31/10/2024 | 149490025,149490010,149490250 |
A0462532 | Certificate of Analysis | 06/05/2024 | 149490025,149490010,149490250 |
A0460631 | Certificate of Analysis | 27/02/2024 | 149490025,149490010,149490250 |
A0459714 | Certificate of Analysis | 29/01/2024 | 149490025,149490010,149490250 |
A0459525 | Certificate of Analysis | 23/01/2024 | 149490025,149490010,149490250 |
Safety and Handling



Product Identifier
- Acetic anhydride
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- EUH071-Corrosive to the respiratory tract.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acetic Anhydride 99+%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction