missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Anhydride, Certified AR for Analysis, Fisher Chemical™
124.60 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
2.5 L
Description
Acetic Anhydride, Certified AR for Analysis
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -73.1°C |
Color | Colorless |
pH | 3 |
Boiling Point | 140°C |
Packaging | HDPE plastic bottle |
Quantity | 2.5 L |
Formula Weight | 102.09g/mol |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'