missing translation for 'onlineSavingsMsg'
Learn More

Acetoin, 96%, may exist as mixture of monomer and dimer, Thermo Scientific Chemicals

£49.95 - £143.72

Chemical Identifiers

CAS 23147-57-1
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00038696
InChI Key DFMGATPNJMFDCR-UHFFFAOYNA-N
Synonym acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl
PubChem CID 179
ChEBI CHEBI:15688
IUPAC Name 3-hydroxybutan-2-one
SMILES CC1OC(C)(O)C(C)OC1(C)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10196840
View Documents Promotion Details
Thermo Scientific Acros
411951000
100 g Glass bottle
£49.95
100g
Please to purchase this item. Need a web account? Register with us today!
10435081
View Documents
Thermo Scientific Acros
411955000
500 g Glass bottle
£143.72
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

23147-57-1
176.21
DFMGATPNJMFDCR-UHFFFAOYNA-N
179
3-hydroxybutan-2-one
C8H16O4
MFCD00038696
acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl
CHEBI:15688
CC1OC(C)(O)C(C)OC1(C)O

Specifications

23147-57-1
100.0
Green-Yellow to Yellow or White to Yellow
148.0°C
Authentic
C8H16O4
1.4171
01, 827
1
Solubility in water: soluble. Other solubilities: soluble in alcohol,propylene glycol,insoluble in vegetable oil
CC1OC(C)(O)C(C)OC1(C)O
176.21
CHEBI:15688
96%
Acetoin, pract., 93%
95.0
15.0°C
1.0000g/mL
50°C
95% min. (GC)
CH3COCH(OH)CH3
MFCD00038696
15, 63
acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl
DFMGATPNJMFDCR-UHFFFAOYNA-N
3-hydroxybutan-2-one
179
88.11
Liquid (Monomer) or Powder or Crystals (Dimer)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 208-174-1

missing translation for 'rtecsNumber' : EL8790000

missing translation for 'tsca' : TSCA

Recommended Storage : may polymerize during storage

Product Identifier
  • Acetoin

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only