Learn More
Acetone, 99.5+%, for analysis, Thermo Scientific Chemicals
£20.40 - £336.68
Chemical Identifiers
CAS | 67-64-1 |
---|---|
Molecular Formula | C3H6O |
Molecular Weight (g/mol) | 58.08 |
MDL Number | MFCD00008765 |
InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
Synonym | acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone |
PubChem CID | 180 |
ChEBI | CHEBI:15347 |
IUPAC Name | propan-2-one |
SMILES | CC(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10295940
|
Thermo Scientific Acros
176805000 |
500 mL | Glass Bottle |
£20.40
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10134100
|
Thermo Scientific Acros
176800010 |
1 L | EcoSafPak™ Glass Bottle |
£30.65
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10194140
|
Thermo Scientific Acros
176800025 |
2.5 L | Glass Bottle |
£62.41
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13188453
|
Thermo Scientific Acros
176800026 |
2.5 L | Plastic Bottle |
£63.01
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10109132
|
Thermo Scientific Acros
176800050 |
5 L | Plastic Drum |
£108.54
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10097962
|
Thermo Scientific Acros
176800250 |
25 L | Metal Drum |
£336.68
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisChemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Specifications
-95.0°C | |
99.5 | |
>4°C | |
56.0°C | |
Liquid | |
99.5+% | |
0.79 | |
CH3COCH3 | |
Vapor Pressure: 247mbar at 20°C | |
01, 635 | |
02,13; 03,4; 06,9 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08 | |
Analytical |
67-64-1 | |
100.0 | |
0.7900g/mL | |
−20°C | |
1.3585 | |
0.001% max. | |
C3H6O | |
MFCD00008765 | |
0.32 mPa.s (20°C) | |
0.5% max. (Coulometric) | |
15, 65 | |
Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
≥99.5% | |
Acetone, For analysis |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Acetone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.