missing translation for 'onlineSavingsMsg'
Learn More

Acetone, 99.6%, for spectroscopy ACS

Product Code. 10045800 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10045800

Brand: Thermo Scientific Acros 400100025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67-64-1 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95.0°C
Absorbance 0.01 max. at 400nm, 0.02 max. at 350nm, 0.1 max. at 340nm, 1 max. at 330nm
CAS Min % 99.5
CAS Max % 100.0
Decomposition Information >4°C
Density 0.7900g/mL
Boiling Point 56.0°C
Quantity 2.5 L
Flash Point −20°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3585
Assay Percent Range 99.6%
Residue after Evaporation 0.001% max.
Specific Gravity 0.79
Titratable Acid 0.0003 meq/g max.
Linear Formula CH3COCH3
Titratable Base 0.0006 meq/g max.
Vapor Pressure Vapor Pressure: 247mbar at 20°C
Viscosity 0.32 mPa.s (20°C)
Beilstein 01, 635
Water 0.5% max. (K.F.)
Fieser 02,13; 03,4; 06,9
Merck Index 15, 65
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
Formula Weight 58.08
Percent Purity 99.6%
Grade ACS
Chemical Name or Material Acetone, Reagent ACS, Spectro Grade
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.