Learn More
Acetone, Certified AR for analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, HDPE plastic bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C
Brand: Fisher Chemical A/0600/PB17
59.57 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
Description
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, HDPE plastic bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C
Specifications
67-64-1 | |
2.5 L | |
C3H6O | |
58.08g/mol | |
0.32 mPaS at 20°C | |
7 | |
56°C |
Plastic Coated Glass bottle | |
Colorless | |
8765 | |
247mbar at 20°C | |
2 | |
-95°C | |
Liquid |
Safety and Handling
- Acetone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.