Learn More
Acetone, Certified AR for analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, HDPE plastic bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C
Brand: Fisher Chemical A/0600/25
302.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 22
Additional Details : CAS Number : 67-64-1 Weight : 19.75000kg Transport : UN number : 1090 Chem class : 3 Pack group : II
Description
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, HDPE plastic bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C
Specifications
67-64-1 | |
25 L | |
C3H6O | |
58.08g/mol | |
0.32 mPaS at 20°C | |
7 | |
56°C |
Metal can | |
Colorless | |
8765 | |
247mbar at 20°C | |
2 | |
-95°C | |
Liquid |