missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
missing translation for 'unitSize'
1 L
2.5 L
Pack of 4
Pack of 6
Beschreibung
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Spezifikation
Spezifikation
| Melting Point | -94°C |
| Density | 0.791g/mL at 25°C |
| Boiling Point | 56°C (760 mmHg) |
| Quantity | 2.5 L |
| Refractive Index | n20/D 1.359 |
| Linear Formula | CH3COCH3 |
| Vapor Pressure | 184mmHg (20°C) |
| Beilstein | 635680 |
| Formula Weight | 58.08g/mol |
| Percent Purity | ≥99.8% |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Acetone
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.,IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : acetone
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur