missing translation for 'onlineSavingsMsg'
Learn More

Acetone, for Electronic Use (MOS), Fisher Chemical™

£120.00 - £354.00

Specifications

CAS 67-64-1
Color Colorless
Molecular Formula C3H6O
MDL Number 8765
Formula Weight 58.08g/mol
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10590711
View Documents Promotion Details
Fisher Chemical
A/0601/17
2.5 L Amber glass bottle
£120.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10659083
View Documents
Fisher Chemical
A/0601/21
5 L Aluminum can
£157.00
5 L
Please to purchase this item. Need a web account? Register with us today!
10440354
View Documents
Fisher Chemical
A/0601/25
25 L Metal can
£354.00
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

67-64-1
C3H6O
58.08g/mol
0.32 mPaS at 20°C
7
56°C
Colorless
8765
247mbar at 20°C
2
-95°C
Liquid
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, for Electronic Use (MOS), Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.