missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Environmental Grade, 99.5+%, Thermo Scientific Chemicals

Suitable For Pesticide Residue Analysis

£34.47 - £248.24

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11374708
View Documents Promotion Details
Thermo Scientific Alfa Aesar
043053.K2
1 L
£34.47
1 L
Please to purchase this item. Need a web account? Register with us today!
11364708
View Documents
Thermo Scientific Alfa Aesar
043053.K7
4 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11354708
View Documents
Thermo Scientific Alfa Aesar
043053.M6
4 x 4 L
£248.24
16 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Suitable for pesticide residue analysisAcetone is a polar, aprotic solvent used in the synthesis and isolation of both organic and inorganic compounds and complexes. It plays a vital role in Jones oxidation. It is utilized for rinsing laboratory glassware due to its low cost and volatility. It is used in the manufacture of various chemicals such as methyl isobutyl ketone, mesityl oxide, diacetone alcohol, acetic acid, iodoform, bromoform, rayon and photographic films, paint and varnish removers. It is also useful in the synthesis of t-butanesulfinamide, terpenes, thiol-stabilized gold colloids, calixphyrin macrocycles and trispyrazolylborate platinum compounds. It plays an important role in protein crystallization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67-64-1
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
C3H6O
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(C)=O

Specifications

°C to 94°C
Colorless
56°C
465°C (869°F)
1.359
C3H6O
185mm Hg at 20°C
635680
Miscible with water,benzene,alcohol,dimethylformamide,ether,chloroform and most oils.
CC(C)=O
58.08
CHEBI:15347
Environmental
67-64-1
0.791
-17°C (1°F)
Liquid
>99.5%
MFCD00008765
1090; Colorless
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
Acetone
Videos
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from sources of ignition - No Smoking. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. If swallowed seek medical advice immediately and show this container or label.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c

H225-H319-H336-H373

missing translation for 'dotInformation' : DOT Class: 3, Packaging Group: II

missing translation for 'einecsNumber' : 200-662-2

missing translation for 'rtecsNumber' : AL3150000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Acetone

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.