Learn More
Acetone, Extra Pure, SLR, Fisher Chemical™
Brand: Fisher Chemical A/0560/PC25
224.47 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 21
Additional Details : CAS Number : 67-64-1 Weight : 19.75000kg Transport : UN number : 1090 Chem class : 3 Pack group : II
Specifications
67-64-1 | |
25 L | |
C3H6O | |
58.08g/mol | |
0.32 mPaS at 20°C | |
7 | |
56°C |
Plastic container | |
Colorless | |
8765 | |
247mbar at 20°C | |
2 | |
-95°C | |
Liquid |
Safety and Handling
- Acetone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.