Learn More
Acetone, Fluka™
Analytical standard, reference material
Brand: Honeywell Fluka R1591-5ML
Additional Details : CAS Number : 67-64-1 Weight : 0.00400kg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifications
Acetone | |
67-64-1 | |
Colorless | |
58.08 | |
-95°C |
Glass Bottle | |
5 mL | |
CH3COCH3 | |
Analytical Standard | |
55°C to 57°C (1.013 hPa) |