missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Laboratory Reagent, ≥99.5%, Honeywell™

Laboratory Reagent,≥99.5%

£32.24 - £456.47

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O
View More Specs

Products 8
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15651640
View Documents Promotion Details
Honeywell Research Chemicals
179973-1L
1 L Glass Bottle
£32.24
1 L
Please to purchase this item. Need a web account? Register with us today!
15688320
View Documents
Honeywell Research Chemicals
179973-4L
4 L Glass Bottle
£104.48
4 L
Please to purchase this item. Need a web account? Register with us today!
15611650
View Documents
Honeywell Research Chemicals
179973-6X1L
6 x 1 L Glass Bottle
£170.06
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15631640
View Documents
Honeywell Research Chemicals
179973-10L
10 L Stainless Steel Can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15691640
View Documents
Honeywell Research Chemicals
179973-4X4L
4 x 4 L Glass Bottle
£331.16
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15601650
View Documents
Honeywell Research Chemicals
179973-4X5L
4 x 5 L Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15671640
View Documents
Honeywell Research Chemicals
179973-20L
20 L Stainless Steel Can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15681640
View Documents
Honeywell Research Chemicals
179973-25L
25 L Stainless Steel Can
£456.47
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

67-64-1
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
C3H6O
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(C)=O

Specifications

-94°C
Colorless
56°C
−17°C (1.4°F)
n20/D 1.359(lit.)
CH3COCH3
184mmHg (20°C)
635680
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08g/mol
Laboratory Reagent
67-64-1
0.791g/mL at 25°C
0.132
Liquid
C3H6O
MFCD00008765
UN1090
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CC(C)=O
58.08
CHEBI:15347
≥99.5%
Acetone
Videos
Safety and Handling

Safety and Handling

P210-P305 + P351 + P338-P370 + P378-P403 + P235

H225-H319-H336

missing translation for 'dotInformation' : class 3 - PG 2 - Acetone

missing translation for 'einecsNumber' : 200-662-2

missing translation for 'rtecsNumber' : AL3150000

Product Identifier
  • Acetone

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : acetone

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.