missing translation for 'onlineSavingsMsg'
Learn More

Acetone, puriss. p.a. ACS Reagent, Honeywell

Product Code. p-7073722
Click to view available options
Quantity:
1 L
2.5 L
155 kg
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Poly-coated Bottle
Unit Size:
1 L
155kg
2.5 L
This item is not returnable. View return policy

Product Code. 15633670

Brand: Honeywell Riedel-de Haën 322011L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Density 0.791g/mL at 25°C
Quantity 1 L
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Packaging Plastic Bottle
Linear Formula CH3COCH3
Vapor Pressure 184mmHg (20°C)
UN Number 1090
Beilstein 635680
Formula Weight 58.08g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Acetone, Reag. Ph. Eur. reag. ISO
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : acetone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.