missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Technical, Fisher Chemical™

Product Code. p-7072771 Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Unit Size:
2.5 L
25 L
This item is not returnable. View return policy

Product Code. 10263750

Brand: Fisher Chemical A/0520/17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Packaging Amber glass bottle
CAS 67-64-1
Vapor Pressure 247mbar at 20°C
Viscosity 0.32 mPaS at 20°C
Vapor Density 2
Physical Form Liquid
Molecular Formula C3H6O
MDL Number 8765
Color Colorless
pH 7
Melting Point -95°C
Boiling Point 56°C
Quantity 2.5 L
Formula Weight 58.08g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.