missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile with 0.1% formic acid, Honeywell Riedel-de Haen™
Click to view available options
2.5 L
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 4.1 |
Melting Point | -46°C |
CAS | 64-18-6, 75-05-8 |
Color | Colorless |
Molecular Formula | CH2O2 |
UN Number | UN1648 |
InChI Key | BDAGIHXWWSANSR-UHFFFAOYSA-N |
SMILES | OC=O |
IUPAC Name | formic acid |
Molecular Weight (g/mol) | 46.03 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Acetonitrile with 0.1% Formic Acid
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : acetonitrile
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'