missing translation for 'onlineSavingsMsg'
Learn More
Acetonitrile with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade, Thermo Scientific™ Product Code.: 10118464

Acetonitrile with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade, Thermo Scientific™

Product Code. 10118464
2.5 L, Glass Bottle
Click to view available options
Quantity:
500mL
1 L
2.5 L
Packaging:
Glass Bottle
Wide Neck Amber Bottle
This item is not returnable. View return policy

Product Code. 10118464

Brand: Fisher Chemical LS120212

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

0.1% Formic Acid in Acetonitrile (v/v), C2H3N, CAS Number-75-05-8, 64-18-6, formisoton, formylic acid, collo-bueglatt, formira, aminic acid, bilorin, acide formique, myrmicyl, hydrogen carboxylic acid, methanoic acid, 2.5L

0.1% Formic Acid in Acetonitrile (v/v), C2H3N, CAS Number-75-05-8, 64-18-6, acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl, 1L

  • Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
  • Pre-mixed for ease-of-use
  • Low metal ion content to ensure minimal metal contamination
  • Formic acid (FA) is a common mobile phase additive that provides protons for LC/MS analysis
  • FA is also known to improve the peak shapes of the resulting separation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8

Specifications

Grade LC-MS
Item Name 0.1% Formic Acid in Acetonitrile (v/v)
Quantity 2.5 L
Form Liquid
Reagent Type Solvent Blends
Packaging Glass Bottle
LCMS Gradient Suitability Pass Test
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile with 0.1% Formic acid (v/v) for HPLC Gradient analysis
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contains : Acetonitrile, Formicacid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetonitrile with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.