Learn More
Acetonitrile with 0.1% Trifluoroacetic Acid (v/v), Optima™ LC/MS Grade, Thermo Scientific™
£135.00 - £597.98
Chemical Identifiers
CAS | 75-05-8, 76-05-1 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
MDL Number | 4169 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10270455
|
Fisher Chemical
LS121-500 |
500 mL | Glass Bottle |
£135.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10230125
|
Fisher Chemical
LS121-1 |
1 L | Wide Neck Amber Bottle |
£275.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10585635
|
Fisher Chemical
LS121-212 |
2.5 L | Glass Bottle |
£493.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11980379
|
Fisher Chemical
LS121-4 |
4 L | Glass Bottle |
£597.98
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
- Pre-mixed for ease-of-use
- Low metal ion content to ensure minimal metal contamination
- TFA is a common mobile phase additive that can function as an ion pair reagent
Chemical Identifiers
75-05-8, 76-05-1 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
4169 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Specifications
2 | |
≤0.03 AU at 254nm, ≤0.4 AU at 230nm, ≤0.55 AU at 220nm, ≤0.6 AU at 210nm | |
>500°C | |
82°C | |
Liquid | |
≤1ppm | |
CH3CN | |
≤0.01% | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
0.095 to 0.105% (v/v) Trifluoroacetic Acid | |
0.1% Trifluoroacetic Acid in Acetonitrile (v/v), Solvent Blends |
-50°C | |
75-05-8, 76-05-1 | |
0.7810g/cm³ | |
Pass Test | |
≥97.5 % | |
C2H3N | |
4169 | |
15, 9847 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.04 | |
OPTIMA™ LC-MS |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.
NFPA
Health:2
Flammability:3
Instability:0
missing translation for 'dotInformation' : DOT Class 3, : Flammable Liquid
- Acetonitrile with 0.1% TFA (v/v) for HPLC Gradient analysis
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.,Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Acetonitrile, Trifluoroacetic acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.