missing translation for 'onlineSavingsMsg'
Learn More
Acetonitrile, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10363413

Acetonitrile, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10363413
500 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10363413

Brand: Thermo Scientific Acros 364315000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46.0°C
CAS Min % 99.85
CAS Max % 100.0
Decomposition Information >500°C
Density 0.7810g/mL
Boiling Point 81.0°C to 82.0°C
Quantity 500 mL
Flash Point 2°C
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Refractive Index 1.3430 to 1.3450
Assay Percent Range 99.9%
Specific Gravity 0.781
Linear Formula CH3CN
Vapor Pressure Vapor Pressure: 97mbar at 20°C
Viscosity 0.36 mPa.s (20°C)
Beilstein 02, 183
Water 0.005% max. (Coulometric)
Fieser 02,13; 12,450; 13,156; 15,1
Merck Index 15, 69
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
Formula Weight 41.05
Percent Purity 99.9%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material Acetonitrile, water <50 ppm, extra dry over mol. sieve
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2534022 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog Number364315000,364310025,364311000,364310010
Lot Number2524004 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number364315000,364310025,364311000,364310010
Lot Number2498940 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number364315000,364310025,364311000,364310010
Lot Number2524004 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number364315000,364310025,364311000,364310010
Lot Number2400165 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number364315000,364310025,364311000,364310010
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetonitrile, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.