missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, 99+%, for spectroscopy, Thermo Scientific Chemicals

£43.22 - £220.10

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10022080
View Documents Promotion Details
Thermo Scientific Acros
167652500
250 mL Glass Bottle
£43.22
250mL
Please to purchase this item. Need a web account? Register with us today!
10194370
View Documents
Thermo Scientific Acros
167650010
1 L Glass Bottle
£103.52
1 L
Please to purchase this item. Need a web account? Register with us today!
10053763
View Documents
Thermo Scientific Acros
167650025
2.5 L Glass Bottle
£220.10
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
Specifications

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N

Specifications

-46.0°C
75-05-8
100.0
0.7810g/mL
2°C
1.3430 to 1.3450
0.0003% max.
C2H3N
MFCD00001878
0.36 mPa.s (20°C)
0.03% max.
15, 69
Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
CC#N
41.053
CHEBI:38472
≥99%
Acetonitrile
0.01 max. at 250nm, 0.02 max. at 235nm, 0.1 max. at 220nm, 0.3 max. at 210nm, 1 max. at 193nm
99.0
>500°C
81.0°C to 82.0°C
Liquid
99+%
0.781
CH3CN
Vapor Pressure: 97mbar at 20°C
02, 183
02,13; 12,450; 13,156; 15,1
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
Spectroscopy
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Product Identifier
  • Acetonitrile

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.