missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile ACS AR, J.T. Baker™
324.90 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle
Description
- For use in Gas Chromatography (GC), Organic Residue Analysis; For Laboratory, Research, or Manufacturing Use
Specifications
CAS | 75-05-8 |
Quantity | 1 L |
Packaging | Narrow Mouth Amber Glass Bottle |
Molecular Formula | C2H3N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
SMILES | CC#N |
IUPAC Name | acetonitrile |
Molecular Weight (g/mol) | 41.053 |
PubChem CID | 6342 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Acetonitrile
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'