missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, AMD CHROMASOLV™, ≥99.9%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
6 x 1 L
Packaging:
Glass Bottle
Description
- Solvents designed for high-performance thin-layer chromatography
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -48°C (literature) |
CAS Max % | 100.0000% |
Density | 0.786 g/mL at 25 °C(lit.) |
Boiling Point | 81.6°C |
Quantity | 1 L |
Flash Point | 6°C |
Packaging | Glass Bottle |
Linear Formula | CH3CN |
UN Number | UN1648 |
Beilstein | 741857 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'