Learn More
Acetonitrile, anhydrous, 99.8+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
£54.57 - £131.66
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
MDL Number | MFCD00001878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15454895
|
Thermo Scientific Alfa Aesar
043166.AK |
250 mL |
£54.57
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15464895
|
Thermo Scientific Alfa Aesar
|
1 L |
£131.66
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acetonitrile is utilized as a polar aprotic solvent in organic synthesis, and in the purification of butadiene. It is used in all the analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS). It is used for the extraction of fatty acids, spinning fibers and casting and molding of plastic materials. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins. It acts as a stabilizer for the chlorinated solvents, and finds use in the production of DNA oligonucleotides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Specifications
°C to 48 to°C to 45°C | |
0.786 | |
5°C (41°F) | |
1.344 | |
C2H3N | |
741857 | |
Miscible with water,alcohols,ethers,acetone,chloroform,carbon tetrachloride and ethylene chloride. Immiscible with saturated hydrocarbons (petroleum fractions). | |
CC#N | |
41.053 | |
CHEBI:38472 | |
Acetonitrile, anhydrous, Water <100ppm |
75-05-8 | |
80°C to 82°C | |
Liquid | |
>99.8% | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 |
Safety and Handling
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c
H225-H302+H312+H332-H319
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II
missing translation for 'einecsNumber' : 200-835-2
missing translation for 'rtecsNumber' : AL7700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Acetonitrile
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.