missing translation for 'onlineSavingsMsg'
Learn More
Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals Product Code.: 11394358

Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals

Product Code. 11394358
250 mL
Click to view available options
Quantity:
250 mL
1 L
This item is not returnable. View return policy

Product Code. 11394358

Brand: Thermo Scientific Alfa Aesar 042311.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Water <100ppm, typically 50ppm (0.005%)

Acetonitrile is utilized as a polar aprotic solvent in organic synthesis, and in the purification of butadiene. It is used in all the analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS). It is used for the extraction of fatty acids, spinning fibers and casting and molding of plastic materials. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins. It acts as a stabilizer for the chlorinated solvents, and finds use in the production of DNA oligonucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecnShow MoreShow Less
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C / -50.8°F
Color Colorless
Appearance Colorless liquid
Density 0.786 g/cm3 at 20°C
Boiling Point 80°C to 82°C
Quantity 250 mL
Flash Point 5°C (41°F)
Ignition Point 525°C (977°F)
Physical Form Liquid
Refractive Index 1.344
Assay Percent Range >99.8%
Vapor Pressure 97 hPa (73mm Hg) at 20°C
UN Number 1648; Colorless
Beilstein 741857
Solubility Information Miscible with water,alcohols,ethers,acetone,chloroform,carbon tetrachloride and ethylene chloride. Immiscible with saturated hydrocarbons (petroleum fractions).
Formula Weight 41.05
Chemical Name or Material Acetonitrile
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX04K967 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number042311.AK,042311.K2,042311.K7
Lot NumberVC740453 Certificate TypeCertificate of Analysis Date03/09/2024 Catalog Number042311.AK,042311.K2,042311.K7
Lot NumberXZ749423 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number042311.AK,042311.K2,042311.K7
Lot NumberR30K829 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number042311.AK,042311.K2,042311.K7
Lot NumberZ04J869 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number042311.AK,042311.K2,042311.K7
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : acetonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.