missing translation for 'onlineSavingsMsg'
Learn More
Acetonitrile Anhydrous (DNA Synthesis), Fisher BioReagents Product Code.: 10428733

Acetonitrile Anhydrous (DNA Synthesis), Fisher BioReagents

Product Code. 10428733
4 L, Glass Bottle
Click to view available options
Quantity:
450 mL
4 L
Packaging:
Amber Glass
Glass Bottle
This item is not returnable. View return policy

Product Code. 10428733

Brand: Fisher BioReagents BP11704

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Acetonitrile Anhydrous is commonly used in DNA or peptide synthesis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecnShow MoreShow Less
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Absorbance 1 max. at 190nm, 0.15 max. at 195nm, 0.07 max. at 200nm, 0.05 max. at 205nm, 0.04 max. at 210nm, 0.02 max. at 220nm, 0.001 max. at 254nm
Color Colorless
Decomposition Information >500°C
Density 0.781g/cm³
Boiling Point 81°C
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3405 to 1.3425 (at 25°C)
Assay Percent Range ≥99.9 %
Residue after Evaporation 1.0ppm max.
Titratable Acid 0.008mEq/g max.
Titratable Base 0.0006mEq/g max.
Vapor Pressure 97mbar at 20°C
UN Number 1648
Viscosity 0.36 cP at 20°C
Water 10ppm max.
Formula Weight 41.05
Percent Purity ≥99.9%
Grade DNA Synthesis
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetonitrile Anhydrous (DNA Synthesis), Fisher BioReagents >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.