missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, BAKER ANALYZED Pesticide Reagent, J.T. Baker™
467.40 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
Description
J.T. Baker BAKER ANALYZED Pesticide Reagent Solvents:
- Suitable for analysis of most common pesticides, such as 2-chlorobiphenyl , Heptachlor, Aldrin, Parathion, Dieldrin, Endrin, DDT, Dioxine, etc.
- Specified to the required low residue levels (5 ppm) for use in research and quality control laboratories
- Ideal for gas chromatography applications, testing for ECD and/or FID sensitive impurities, organic residues, and residue after evaporation
Specifications
Melting Point | -46°C |
CAS | 75-05-8 |
Color | Colorless |
Acidity | Max. 0.0005meq/g |
Molecular Formula | C2H3N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
Solubility Information | Fully miscible in water |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
SMILES | CC#N |
IUPAC Name | acetonitrile |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Acetonitrile
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'