missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Certified AR for Analysis, Fisher Chemical™

C2H3N, CAS Number-75-05-8, 5L, 81 deg.C, Colorless, 41.05g/mol, -46 deg.C, 1878, Metal can, Liquid, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C

£176.00 - £1910.00

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
MDL Number 1878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10427440
View Documents
Fisher Chemical
A/0621/PB15
1 L Plastic-coated Glass Bottle
£176.00
1 L
Please to purchase this item. Need a web account? Register with us today!
10345070
View Documents Promotion Details
Fisher Chemical
A/0621/PB17
2.5 L Plastic-coated Glass Bottle
£335.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10253800
View Documents
Fisher Chemical
A/0621/21
5 L Metal Can
£480.00
5 L
Please to purchase this item. Need a web account? Register with us today!
10764361
View Documents
Fisher Chemical
A/0621/25
25 L Metal Can
£1910.00
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

C2H3N, CAS Number-75-05-8, 5L, 81 deg.C, Colorless, 41.05g/mol, -46 deg.C, 1878, Metal can, Liquid, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C

Specifications

Chemical Identifiers

75-05-8
41.05
WEVYAHXRMPXWCK-UHFFFAOYSA-N
CC#N
C2H3N
1878
acetonitrile

Specifications

-46°C
Colorless
Liquid
1878
0.36cp at 20°C
CC#N
41.05
75-05-8
81°C
C2H3N
97mbar at 20°C
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
41.05g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Acetonitrile

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

SDS
Documents

Documents

Product Certifications