missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, ChromAR™, Macron Fine Chemicals™

Product Code. 15534065
Click to view available options
Quantity:
2.5 L
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Stored in an amber, narrow mouth glass bottle for protection from light. Macron Fine Chemicals™ ChromAR™ Acetonitrile meets ACS specifications and is suitable for liquid chromatography and UV-spectrophotometry.

Specifications

Melting Point −46°C
Absorbance Max. 1.00 at 190nm, Max. 0.05 at 200-210nm, Max. 0.02 at 220nm, Max. 0.01 at 230nm, Max. 0.005 at 254-400nm
CAS 75-05-8
Color Clear, colorless
Appearance Passes Test
Density 0.79g/mL at 20°C
Boiling Point 81°C
Quantity 2.5 L
Explosion Limit Lower: 4.4% Vol, Upper: 16% Vol
Flash Point 2.00°C
Ignition Point 524°C
Odor Ether-like
Packaging Narrow Mouth Amber Glass Bottle
Physical Form Liquid
Residue after Evaporation Max. 0.0002%
Molecular Formula CH3CN
Titratable Acid Max. 8μeq/g
Titratable Base Max. 0.6μeq/g
Vapor Pressure 9.73 kPa at 20°C
Water Max. 0.02%, by Karl Fischer titration
Molecular Weight (g/mol) 41.05
Percent Purity ≥99.8%
Grade ChromAR™
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'