missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën Acetonitrile CHROMASOLV™

Product Code. 15935480
Click to view available options
Quantity:
7 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Acetonitrile CHROMASOLV™ for pesticide residue analysis, ≥99.9% (GC).

  • For gas chromatography (GC).
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC.
  • Non-volatile matter and water content are specified for each solvent.

Specifications

pH Not applicable
Melting Point -46°C
CAS 75-05-8
Color Colorless
CAS Max % 100.00%
Density 0.7857 g/cm3 (20°C)
Boiling Point 82°C (1.013 hPa)
Quantity 7 L
Flash Point 6°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line CHROMASOLV™
Molecular Formula C2H3N
Linear Formula CH3CN
MDL Number MFCD00001878
Vapor Pressure 97 hPa (20°C)
UN Number UN1648
Viscosity No data available
Beilstein 741857
Synonym ACN, Methyl cyanide
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.05
Percent Purity ≥99.90%
Grade GC
Chemical Name or Material Acetonitrile
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'