missing translation for 'onlineSavingsMsg'
Learn More
Riedel-de Haën Acetonitrile CHROMASOLV™ Product Code.: 15616896
compliance-icons compliance-icons

Riedel-de Haën Acetonitrile CHROMASOLV™

Product Code. 15616896
7 L
Click to view available options
Quantity:
7 L
18 L
45 L
This item is not returnable. View return policy

Product Code. 15616896

Brand: Riedel-de Haën 348517L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Acetonitrile CHROMASOLV™, gradient grade, for HPLC, ≥99.9%.

  • Same high quality as CHROMASOLV HPLC.
  • Additional testing for HPLC via gradient analysis and UV-VIS spectroscopy.
  • Low UV profile by HPLC.
  • Low impurity gradient LC baseline.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym ACN, Methyl cyanide
IUPAC Name acetonitrile
SMILES CC#N

Specifications

pH Not applicable
Melting Point -46°C
Color Colorless
CAS Max % 100.00%
Density 0.7857 g/cm3 (20°C)
Boiling Point 82°C (1.013 hPa)
Quantity 7 L
Flash Point 6°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line CHROMASOLV™
Linear Formula CH3CN
Vapor Pressure 97 hPa (20°C)
UN Number UN1648
Viscosity No data available
Beilstein 741857
Concentration 100.00%
Percent Purity ≥99.90%
Grade HPLC Gradient
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Riedel-de Haën Acetonitrile CHROMASOLV™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.