missing translation for 'onlineSavingsMsg'
Learn More
Acetonitrile, CHROMASOLV™, ≥99.9% (GC), for LC-NMR, Honeywell Riedel-de Haën™ Product Code.: 15634720

Acetonitrile, CHROMASOLV™, ≥99.9% (GC), for LC-NMR, Honeywell Riedel-de Haën™

Product Code. 15634720
6 x 1 L
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
6 x 1 L
This item is not returnable. View return policy

Product Code. 15634720

Brand: Honeywell-Chromasolv 349556X1L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHROMASOLV™ NMR, for LC-NMR,≥99.9% (GC)

  • Solvents for LC-NMR applications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecnShow MoreShow Less
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Color Colorless
Density 0.786g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Linear Formula CH3CN
Vapor Pressure 72.8 mmHg (20°C)
UN Number UN1648
Formula Weight 41.05
Percent Purity ≥99.9% (GC)
Chemical Name or Material Acetonitrile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetonitrile, CHROMASOLV™, ≥99.9% (GC), for LC-NMR, Honeywell Riedel-de Haën™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.