missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, CHROMASOLV™, for HPLC, gradient grade, ≥99.9%, Honeywell Riedel-de Haën™
86.94 GBP valid until 2024-12-31
Use promo code "21944" to get your promotional price.
Use promo code "21944" to get your promotional price.
Click to view available options
Quantity:
1 L
100 mL
2.5 L
2.5 L-PC
4 x 2.5 L
6 x 1 L
Description
- Same high quality as CHROMASOLV HPLC
- Additional testing for HPLC via gradient analysis and UV-VIS spectroscopy
- Low UV profile by HPLC
- Low impurity gradient LC baseline
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -48°C |
Absorbance | HPLC-gradient/254nm ≤0.5mAU |
Density | 0.786g/mL at 25°C |
Boiling Point | 81°C to 82°C |
Quantity | 1 L |
Refractive Index | n20/D 1.344 |
Linear Formula | CH3CN |
Vapor Pressure | 72.8mmHg (20°C) |
Beilstein | 741857 |
Formula Weight | 41.05g/mol |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Acetonitrile
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'