missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, CHROMASOLV™, for HPLC, gradient grade, ≥99.9%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
Specifications
Specifications
CAS Max % | 100.0000% |
UN Number | UN1648 |
Beilstein | 741857 |
Percent Purity | >99.9% |
Grade | HPLC |
Quantity | 4 x 2.5 L |
Packaging | Glass Bottle |
Chemical Name or Material | Acetonitrile, CHROMASOLV, for HPLC, gradient grade, 99.9% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'